Tisk…

Potassium cyclopropyltrifluoroborate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

662984-1G 662984-5G
SIAL662984-1GEA 2950 CZK
SIAL662984-1G SIAL662984-5G
Potassium cyclopropyltrifluoroborate, Sigma-Aldrich®
Potassium cyclopropyltrifluoroborate
Vzorec: C₃H₅BF₃K
MDL Number: MFCD09265031
CAS číslo: 1065010-87-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR