3-Phenyl-1-propyne, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Propynylbenzene
M-Clarity™ quality level = MQ100
376841-1G
376841-5G
SIAL376841-1GEA
2830
CZK
SIAL376841-1G
SIAL376841-5G
3-Phenyl-1-propyne, Sigma-Aldrich®
3-Phenyl-1-propyne
Vzorec:
C₉H₈ MW: 116,16 g/mol |
MDL Number:
MFCD00134431 CAS číslo: 10147-11-2 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...