Tisk…
Perfluoroheptanoic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Tridecafluoroenanthic acid, Tridecafluoroheptylic acid, Tridecafluoroheptanoic acid, Perfluoroenanthic acid
M-Clarity™ quality level = MQ200
342041-25G 342041-5G
SIAL342041-25GEA 4670 CZK
SIAL342041-25G SIAL342041-5G
Perfluoroheptanoic acid, Sigma-Aldrich®
Perfluoroheptanoic acid
Vzorec: C₇HF₁₃O₂
MDL Number: MFCD00039604
CAS číslo: 375-85-9
EINECS: 206-798-9
UN: 3261
ADR: 8,III