Tisk…

Perfluoro-tert-butanol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Perfluoro-t-butanol, Nonafluoro-tert-butyl alcohol, 1,1,1,3,3,3-Hexafluoro-2-trifluoromethyl-2-propanol, Perfluoro-tert-butyl alcohol

M-Clarity™ quality level = MQ200

331023-1G 331023-5G
SIAL331023-1GEA 2540 CZK
SIAL331023-1G SIAL331023-5G
Perfluoro-tert-butanol, Sigma-Aldrich®
Perfluoro-tert-butanol
Vzorec: C₄HF₉O
MW: 236,04 g/mol
Bod varu: 84 °C (1013 hPa)
Bod tání: –17 °C
Hustota: 1,693 g/cm³ (20 °C)
MDL Number: MFCD00042092
CAS číslo: 2378-02-1
EINECS: 219-157-3
UN: 2810
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR