Tisk…
Cobalt(II) thiocyanate, Sigma-Aldrich®
Dodavatel: Merck
Varování
Synonyms: Cobalt dirhodanide, Cobaltous rhodanide, Cobalt(II) thiocyanate, Cobalt dithiocyanate, Cobaltous thiocyanate
M-Clarity™ quality level = MQ100
216135-125G 216135-25G 216135-5G
SIAL216135-125GEA 20010 CZK
SIAL216135-125G SIAL216135-25G SIAL216135-5G
Cobalt(II) thiocyanate, Sigma-Aldrich®
Cobalt(II) thiocyanate
Vzorec: C₂CoN₂S₂
MW: 175.1
Hustota: 1.955 g/cm³ (25 °C)
MDL Number: MFCD00010937
CAS číslo: 3017-60-5
EINECS: 221-156-8