Tisk…

Propargyl chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Chloropropyne, 3-Chloro-1-propyne

M-Clarity™ quality level = MQ100

143995-25G 143995-5G
SIAL143995-25GEA 3150 CZK
SIAL143995-25G SIAL143995-5G
Propargyl chloride, Sigma-Aldrich®
Propargyl chloride
Vzorec: C₃H₃Cl
MW: 74.51
Bod varu: 58
Bod tání: –77 °C
Hustota: 1.0454 g/cm³ (5 °C)
MDL Number: MFCD00000980
CAS číslo: 624-65-7
EINECS: 210-856-9
UN: 3286
ADR: 3 I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR