Tisk…

1,5-Hexadien-3-ol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Hexa-1,5-dien-3-ol

M-Clarity™ quality level = MQ100

118559-10G 118559-25G
SIAL118559-10GEA 9320 CZK
SIAL118559-10G SIAL118559-25G
1,5-Hexadien-3-ol, Sigma-Aldrich®
1,5-Hexadien-3-ol
Vzorec: C₆H₁₀O
MW: 98,14 g/mol
MDL Number: MFCD00008662
CAS číslo: 924-41-4
EINECS: 213-102-7
UN: 1987
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR