Potassium (3-bromophenyl)trifluoroborate ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
potassium (3-bromophenyl)trifluoroborate, (3-Bromophenyl)trifluoroboric acid potassium salt
L17966.03
L17966.06
L17966.03EA
831
CZK
L17966.03
L17966.06
Potassium (3-bromophenyl)trifluoroborate ≥97%
Potassium (3-bromophenyl)trifluoroborate
Purity: 97%
CAS No.: 374564-34-8
Molecular Formula: C6H4BBrF3K
Formula Weight: 262.91
Melting Point: 178-182°
MDL No.: MFCD02093977
Vzorec:
C₆H₄BBrF₃K MW: 262,91 g/mol Bod tání: 178…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093977 CAS číslo: 374564-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...