Potassium phenyltrifluoroborate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L17568.03
L17568.06
L17568.03EA
1180
CZK
L17568.03
L17568.06
Potassium phenyltrifluoroborate ≥98%
Potassium phenyltrifluoroborate
Purity: 98%
CAS No.: 153766-81-5
Molecular Formula: C6H5BF3K
Formula Weight: 184.00
Melting Point: 296-301°
Beilstein Reference: 7782070
Vzorec:
C₆H₅BF₃K Bod tání: 296…301 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318172 CAS číslo: 153766-81-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...