1H,1H-Nonadecafluoro-1-decanol ≥98%
Dodavatel: Thermo Fisher Scientific
L16580.06
L16580.14
L16580.06EA
2160
CZK
L16580.06
L16580.14
1H,1H-Nonadecafluoro-1-decanol ≥98%
1H,1H-Nonadecafluoro-1-decanol
Purity: 98%
CAS No.: 307-37-9
Molecular Formula: C10H3F19O
Formula Weight: 500.10
Melting Point: 83-90°
Boiling Point: 95°/20mm
MDL No.: MFCD00153234
Vzorec:
C₁₀H₃F₁₉O Bod varu: 95 °C (20 mmHg) Bod tání: 83…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153234 CAS číslo: 307-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...