Tisk…

Potassium (4-iodophenyl)trifluoroborate 96%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (4-Iodophenyl)trifluoroboric acid potassium salt, Potassium 4-iodophenyltrifluoroborate

H28758.06 H28758.03
H28758.06EA 7690 CZK
H28758.06 H28758.03
Potassium (4-iodophenyl)trifluoroborate 96%
Potassium (4-iodophenyl)trifluoroborate

Purity: 96% CAS No.: 912350-00-6 Molecular Formula: C6H4BF3IK Formula Weight: 309.90 Melting Point: 297-299° MDL No.: MFCD09800738

Vzorec: C₆H₄BF₃IK
MW: 309.91
Bod tání: 297…299 °C
Storage Temperature: Ambient
MDL Number: MFCD09800738
CAS číslo: 912350-00-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR