ω-Bromo-2-hydroxyacetophenone ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Hydroxyphenacyl bromide, 2-Bromo-1-(2-hydroxyphenyl)ethan-1-one, 2-Bromo-1-(2-hydroxyphenyl)ethanone, α-Bromo-2-hydroxyacetophenone, 2-Bromo-2'-hydroxyacetophenone
B25722.06
B25722.14
B25722.06EA
2850
CZK
B25722.06
B25722.14
ω-Bromo-2-hydroxyacetophenone ≥97%
ω-Bromo-2-hydroxyacetophenone
Purity: 97%
CAS No.: 2491-36-3
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
Melting Point: 45-47°
UN No.: 3263
MDL No.: MFCD01727570
Vzorec:
C₈H₇BrO₂ MW: 215,05 g/mol Bod tání: 45…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD01727570 CAS číslo: 2491-36-3 UN: 3263 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...