2'-Hydroxy-4',6'-dimethoxyacetophenone ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-(2-Hydroxy-4,6-dimethoxyphenyl)ethan-1-one, Xanthoxylin
A14585.03
A14585.06
A14585.03EA
1660
CZK
A14585.03
A14585.06
2'-Hydroxy-4',6'-dimethoxyacetophenone ≥98%
2'-Hydroxy-4',6'-dimethoxyacetophenone
Purity: 98%
CAS No.: 90-24-4
Molecular Formula: C10H12O4
Formula Weight: 196.20
Melting Point: 80-83°
Boiling Point: 185°/20mm
Merck Reference: 14,10067
MDL No.: MFCD00017243
Vzorec:
C₁₀H₁₂O₄ MW: 196,2 g/mol Bod varu: 185 °C (20 mmHg) Bod tání: 80…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017243 CAS číslo: 90-24-4 EINECS: 201-978-3 Merck Index: 14,10067 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...