Tisk…

4-Bromo-2-methyl-6-nitroaniline ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4-Bromo-6-nitro-o-toluidine, 2-Amino-5-bromo-3-nitrotoluene

A14319.14 A14319.06
A14319.14EA 7730 CZK
A14319.14 A14319.06
4-Bromo-2-methyl-6-nitroaniline ≥97%
4-Bromo-2-methyl-6-nitroaniline

Purity: 97% CAS No.: 77811-44-0 Molecular Formula: C7H7BrN2O2 Formula Weight: 231.06 Melting Point: 143-147° Beilstein Reference: 3530524

Vzorec: C₇H₇BrN₂O₂
MW: 231,05 g/mol
Bod tání: 143…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00052919
CAS číslo: 77811-44-0
UN: 3288
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR