Propiolaldehyde diethyl acetal 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Propargylaldehyde diethyl acetal, 3,3-Diethoxypropyne, Propioaldehyde diethyl acetal, Propynal diethyl acetal, 3,3-Diethoxy-1-propyne, 3,3-Diethoxyprop-1-yne
382920010
382920050
ACRO382920010EA
758
CZK
ACRO382920010
ACRO382920050
Propiolaldehyde diethyl acetal 98%
Propiolaldehyde diethyl acetal
Vzorec:
C₇H₁₂O₂ MW: 128.17 Bod varu: 137…139 °C (760 mmHg) Hustota: 0.894 g/cm³ Bod vzplanutí: 31 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009237 CAS číslo: 10160-87-9 EINECS: 233-430-4 UN: 1993 ADR: 3 III |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4110 to 1.4140 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...