Propagyl amine hydrochloride 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Propargylamine hydrochloride, Prop-2-yn-1-amine hydrochloride
131470010
131470100
ACRO131470010EA
1460
CZK
ACRO131470010
ACRO131470100
Propagyl amine hydrochloride 95%
Propagyl amine hydrochloride
Vzorec:
C₃H₆ClN MW: 91,54 g/mol Bod tání: 175…182 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00012907 CAS číslo: 15430-52-1 EINECS: 239-440-5 UN: 2811 ADR: 6.1,III |
Výsledky specifikačního testu
Appearance | White to beige to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 175 to 182 °C |
Titration Argentometric | 94 to 106 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...