Tisk…

n-Octacosane ≥99%

Dodavatel: Thermo Fisher Scientific

42460.09 42460.18 42460.30
42460.09EA 1030 CZK
42460.09 42460.18 42460.30
n-Octacosane ≥99%
n-Octacosane

Purity: 99% CAS No.: 630-02-4 Molecular Formula: CH3(CH2)26CH3 Formula Weight: 394.77 Flash Point: 227°(441°F) Physical Form: Crystalline Melting Point: 61-63° Boiling Point: 440° MDL No.: MFCD00009355

Vzorec: H₃C(CH₂)₂₆CH₃
Bod varu: 440 °C
Bod tání: 61…63 °C
Hustota: 0,8067 g/cm³ (20 °C)
Bod vzplanutí: 227 °C (441 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009355
CAS číslo: 630-02-4
EINECS: 211-125-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR