Tisk…

n-Nonadecane, Supelco®

Dodavatel: Merck

Synonyms: N-nonadecane

MQ100

74158-1G 74158-5G
74158-1GEA 808 CZK
74158-1G 74158-5G
n-Nonadecane, Supelco®
n-Nonadecane
Vzorec: H₃C(CH₂)₁₇CH₃
MW: 268.53
Bod varu: 329.7
Bod tání: 32.5
Hustota: 0.7845 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009012
CAS číslo: 629-92-5
EINECS: 211-116-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b04de4ba35a4af2a5beef17009e6a5176edae15b&vid=6891660a5fcf1a7cf224756156004b5284d3e169&prid=web&clid=SAPDAM

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR