Tisk…

n-Heptacosane ≥97%

Dodavatel: Thermo Fisher Scientific

Synonyms: Heptacosane

L07796.03 L07796.MD
L07796.03EA 5430 CZK
L07796.03 L07796.MD
n-Heptacosane ≥97%
n-Heptacosane

Purity: 97% CAS No.: 593-49-7 Molecular Formula: CH3(CH2)25CH3 Formula Weight: 380.74 Flash Point: 120°(248°F) Melting Point: 57-60° Boiling Point: 270°/15mm Beilstein Reference: 1706242 MDL No.: MFCD00009862

Vzorec: H₃C(CH₂)₂₅CH₃
MW: 380.74
Bod varu: 270 °C (15 mmHg)
Bod tání: 57…60 °C
Hustota: 0.7796 g/cm³ (60 °C)
Bod vzplanutí: 120 °C (248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009862
CAS číslo: 593-49-7
EINECS: 209-792-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR