n-Dotriacontane ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Dotriacontane
A17480.06
A17480.14
A17480.06EA
1390
CZK
A17480.06
A17480.14
n-Dotriacontane ≥98%
n-Dotriacontane
Purity: 98%
CAS No.: 544-85-4
Molecular Formula: CH3(CH2)30CH3
Formula Weight: 450.88
Melting Point: 68-71°
Boiling Point: 300-310°
MDL No.: MFCD00009411
Vzorec:
H₃C(CH₂)₃₀CH₃ MW: 450.88 Bod varu: 300…310 °C Bod tání: 68…71 °C Hustota: 0.812 Storage Temperature: Ambient |
MDL Number:
MFCD00009411 CAS číslo: 544-85-4 EINECS: 208-881-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...