Tisk…

n-Dotriacontane 97%

Dodavatel: Thermo Fisher Scientific

Synonyms: Dotriacontane

117690250 117690050 117691000
ACRO117690250EA 4310 CZK
ACRO117690250 ACRO117690050 ACRO117691000
n-Dotriacontane 97%
n-Dotriacontane
Vzorec: H₃C(CH₂)₃₀CH₃
MW: 450.88
Bod varu: 467 °C (760 mmHg)
Bod tání: 67…72 °C
Bod vzplanutí: 247 °C
Storage Temperature: Ambient
MDL Number: MFCD00009411
CAS číslo: 544-85-4
EINECS: 208-881-5

Objednat

Výsledky specifikačního testu

Appearance White to light grey Glistening flakes
Infrared spectrum Conforms
Melting point 67 to 72 °C
GC ≥96.0 %
Clarity of alcohol solution Passes test
Specific optical rotation +52.0° to +62.0° (20 °C, 589 nm) (neat)

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR