1-Pentanethiol ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 97%
CAS No.: 110-66-7
Molecular Formula: CH3(CH2)4SH
Formula Weight: 104.22
Flash Point: 18°(64°F)
Refractive Index: 1.4460
Odour: Stench
Melting Point: -76°
Boiling Point: 125-126°
Beilstein Reference: 1730979
UN No.: 1111
MDL No.: MFCD00004907
Vzorec:
C₅H₁₂S Bod varu: 125…126 °C Bod tání: –76 °C Hustota: 0,841 Bod vzplanutí: 18 °C (64 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004907 CAS číslo: 110-66-7 EINECS: 203-789-1 UN: 1111 ADR: 3,II Merck Index: 12,00653 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...