1-Octadecanethiol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
n-Octadecyl mercaptan, n-Octadecanethiol, Stearyl mercaptane, 1-Mercaptooctadecane, Octadecane-1-thiol
M-Clarity™ quality level = MQ200
74731-250G
74731-50G
SIAL74731-250GEA
11690
CZK
SIAL74731-250G
SIAL74731-50G
1-Octadecanethiol, Sigma-Aldrich®
1-Octadecanethiol
Vzorec:
C₁₈H₃₈S MW: 286,57 g/mol Bod varu: 204 °C (1013 hPa) Bod tání: 28…31 °C Hustota: 0,8475 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004886 CAS číslo: 2885-00-9 EINECS: 220-744-1 UN: 3335 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...