Etylenglykol 99+% for spectrophotometry
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol
Purity: 99+%
CAS No.: 107-21-1
Molecular Formula: C2H6O2
Formula Weight: 62.07
Flash Point: 119°(246°F)
Physical Form: Liquid
Density: 1.113
Refractive Index: 1.4310
Sensitivity: Hygroscopic
Appearance: Colourless
Odour: Sweet
Melting Point: -13°
Boiling Point: 196-198°
Beilstein Reference: 505945
Merck Reference: 14,3798
MDL No.: MFCD00002885
For use in spectrophotometry
Vzorec:
HOCH₂CH₂OH MW: 62.07 Bod varu: 196…198 °C Bod tání: –13 °C Hustota: 1.113 Bod vzplanutí: 119 °C (246 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002885 CAS číslo: 107-21-1 EINECS: 203-473-3 REACH: 01-2119456816-28 Merck Index: 14,3798 |