(±)-3-Nonanol ≥98%
Dodavatel: Thermo Scientific
Synonyms:
(±)-Hexyl ethyl carbinol, (±)-3-Hydroxy nonane, (±)-Ethyl hexyl carbinol
A19120.18
A19120.30
A19120.18EA
3050
CZK
A19120.18
A19120.30
(±)-3-Nonanol ≥98%
(±)-3-Nonanol
Purity: 98%
CAS No.: 624-51-1
Molecular Formula: C9H20O
Formula Weight: 144.26
Flash Point: 79°(174°F)
Refractive Index: 1.4310
Boiling Point: 192-194°
Beilstein Reference: 1719453
MDL No.: MFCD00014411
Vzorec:
C₉H₂₀O MW: 144,26 g/mol Bod varu: 192…194 °C Bod tání: –23…–22 °C Hustota: 0,824 Bod vzplanutí: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014411 CAS číslo: 624-51-1 EINECS: 210-850-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...