(±)-3-Hexanol ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(±)-Propyl ethyl carbinol, (±)-Ethyl propyl carbinol, (±)-3-Hydroxy hexane
Purity: 98%
CAS No.: 623-37-0
Molecular Formula: C6H14O
Formula Weight: 102.18
Flash Point: 42°(108°F)
Refractive Index: 1.4150
Odour: Characteristic
Boiling Point: 134-136°
Beilstein Reference: 1718964
UN No.: 2282
MDL No.: MFCD00004582
Vzorec:
C₆H₁₄O MW: 102,18 g/mol Bod varu: 134…136 °C Bod tání: –70 °C Hustota: 0,81 Bod vzplanutí: 42 °C (108 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004582 CAS číslo: 623-37-0 EINECS: 210-790-0 UN: 2282 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...