(±)-2-Decanol ≥98%
Dodavatel: Thermo Scientific
Varování
|
Synonyms:
(±)-Methyl octyl carbinol, (±)-2-Hydroxy decane, 2-Decanol, (±)-Octyl methyl carbinol
Purity: 98%
CAS No.: 1120-06-5
Molecular Formula: C10H22O
Formula Weight: 158.29
Flash Point: 85°(185°F)
Refractive Index: 1.4340
Melting Point: -5°
Boiling Point: 211°
Beilstein Reference: 1719664
MDL No.: MFCD00004594
Vzorec:
C₁₀H₂₂O MW: 158,28 g/mol Bod varu: 211 °C Bod tání: –5 °C Hustota: 0,825 Bod vzplanutí: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004594 CAS číslo: 1120-06-5 EINECS: 214-296-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...