1-Propanol, anhydrous 99,9
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane
Argon_Chemseal
Purity: 99.9%
CAS No.: 71-23-8
Molecular Formula: CH3CH2CH2OH
Formula Weight: 60.10
Flash Point: 15°(59°F)
Physical Form: Liquid
Density: 0.804
Refractive Index: 1.3850
Odour: Alcohol-like
Melting Point: -127°
Boiling Point: 97°
Beilstein Reference: 1098242
Merck Reference: 14,7842
UN No.: 1274
MDL No.: MFCD00002941
Vzorec:
H₃CCH₂CH₂OH MW: 60.1 Bod varu: 97 °C Bod tání: –127 °C Hustota: 0.804 Bod vzplanutí: 15 °C (59 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002941 CAS číslo: 71-23-8 EINECS: 200-746-9 UN: 1274 ADR: 3 II Merck Index: 14,7842 |