Tisk…

Arachidyl alcohol 96%

Dodavatel: Thermo Scientific

Synonyms: 1-Eicosanol

L08807.14 L08807.22
L08807.14EA 927 CZK
L08807.14 L08807.22
Arachidyl alcohol 96%
Arachidyl alcohol
Vzorec: H₃C(CH₂)₁₈CH₂OH
MW: 298,55 g/mol
Bod varu: 220…225 °C (3 mmHg)
Bod tání: 62…66 °C
Hustota: 0,841
Bod vzplanutí: 195 °C (383 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002938
CAS číslo: 629-96-9
EINECS: 211-119-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR