1-Dodekanol ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Undecyl carbinol, 1-Dodecyl alcohol, 1-Hydroxy dodecane, Lauryl alcohol, Dodecyl Alcohol
Purity: 98%
CAS No.: 112-53-8
Molecular Formula: C12H26O
Formula Weight: 186.34
Flash Point: 127°(260°F)
Physical Form: Crystalline
Density: 0.830
Refractive Index: 1.4430
Appearance: White
Odour: Alcohol-like
Melting Point: 24-27°
Boiling Point: 258-260°
Beilstein Reference: 1738860
Merck Reference: 14,3405
UN No.: 3077
MDL No.: MFCD00004753
Vzorec:
H₃C(CH₂)₁₀CH₂OH MW: 186,34 g/mol Bod varu: 258…260 °C Bod tání: 24…27 °C Hustota: 0,830 Bod vzplanutí: 127 °C (260 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004753 CAS číslo: 112-53-8 EINECS: 203-982-0 UN: 3077 ADR: 9,III Merck Index: 14,3405 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...