1-Butanol, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol
M-Clarity™ quality level = MQ100
281549-100ML
281549-1L
SIAL281549-100MLEA
2760
CZK
SIAL281549-100ML
SIAL281549-1L
1-Butanol, Sigma-Aldrich®
1-Butanol
Vzorec:
H₃C(CH₂)₂CH₂OH MW: 74.12 Bod varu: 117.6 Bod tání: –89 °C Hustota: 0.812 g/cm³ (20 °C) Bod vzplanutí: 30 Storage Temperature: Ambient |
MDL Number:
MFCD00002964 CAS číslo: 71-36-3 EINECS: 200-751-6 UN: 1120 ADR: 3 III REACH: 01-2119484630-38 Merck Index: 13,01539 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...