Tisk…

Formaldehyddimethylacetal ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Formal, Methylal, Dimethoxymethane

A12055.0F A12055.AE A12055.AP AB116047.2500
A12055.0FEA 3160 CZK
A12055.0F A12055.AE A12055.AP ABCRAB116047.2500
Formaldehyddimethylacetal ≥98%
Formaldehyddimethylacetal

Purity: 98% CAS No.: 109-87-5 Molecular Formula: C3H8O2 Formula Weight: 76.10 Flash Point: -17°(1°F) Refractive Index: 1.3545 Melting Point: -105° Boiling Point: 41-42° Beilstein Reference: 1697025 Merck Reference: 14,6012 UN No.: 1234 MDL No.: MFCD00008495

Vzorec: CH₂(OCH₃)₂
MW: 76,1 g/mol
Bod varu: 41…42 °C
Bod tání: –105 °C
Hustota: 0,863
Bod vzplanutí: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008495
CAS číslo: 109-87-5
EINECS: 203-714-2
UN: 1234
ADR: 3,II
Merck Index: 14,6012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7dc068e2803e769f0f748f5152cbff2cbfe69fc&vid=eae965f4719d855b0e2585391d097f07b53b9067&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR