Tisk…

(±)-sec-Butylamin 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: (±)-2-Butylamine, (±)-2-Aminobutane

A13014.0F A13014.AP
A13014.0FEA 5530 CZK
A13014.0F A13014.AP
(±)-sec-Butylamin 98%
(±)-sec-Butylamin

Purity: 98% CAS No.: 13952-84-6 Molecular Formula: C4H11N Formula Weight: 73.14 Flash Point: -19°(-2°F) Refractive Index: 1.3930 Odour: Amine-like pH: 12.5 Melting Point: -104° Boiling Point: 63-64° Beilstein Reference: 1361345 Merck Reference: 14,1544 UN No.: 2733 MDL No.: MFCD00008094

Vzorec: C₄H₁₁N
MW: 73.14
Bod varu: 63…64 °C
Bod tání: –104 °C
Hustota: 0.722
Bod vzplanutí: –19 °C (–2 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008094
CAS číslo: 13952-84-6
EINECS: 237-732-7
UN: 2733
ADR: 3 II
Merck Index: 12,01579

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR