Tetradecylamine 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
1-Tetradecylamine, 1-Aminotetradecane, n-Tetradecylamine, Tetradecan-1-amine
138100100
138100050
138100500
ACRO138100100EA
2190
CZK
ACRO138100100
ACRO138100050
ACRO138100500
Tetradecylamine 98%
Tetradecylamine
Vzorec:
H₃C(CH₂)₁₂CH₂NH₂ MW: 213.41 Bod varu: 291 °C (760 mmHg) Bod tání: 37…42 °C Hustota: 0.8571 Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008157 CAS číslo: 2016-42-4 EINECS: 217-950-9 UN: 3259 ADR: 8 III |
Výsledky specifikačního testu
Appearance | White to pink Crystals or crystalline solid |
Infrared spectrum | Conforms |
Melting point | 37 to 42 °C |
Titration with HClO4 | ≥97.5 % |
Elemental Analysis | 48.8 to 59.6 % (O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...