Tisk…

Propylamin 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1-Propylamine, 1-Aminopropane

A11606.0F A11606.AP
A11606.0FEA 3470 CZK
A11606.0F A11606.AP
Propylamin 98%
Propylamin

Purity: 98% CAS No.: 107-10-8 Molecular Formula: CH3CH2CH2NH2 Formula Weight: 59.11 Flash Point: -12°(10°F) Refractive Index: 1.389 Odour: Amine-like pH: 12.6 Melting Point: -83° Boiling Point: 48-49° Beilstein Reference: 1098243 Merck Reference: 14,7843 UN No.: 1277 MDL No.: MFCD00008205

Vzorec: H₃CCH₂CH₂NH₂
MW: 59,11 g/mol
Bod varu: 48…49 °C
Bod tání: –83 °C
Hustota: 0,719
Bod vzplanutí: –12 °C (10 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008205
CAS číslo: 107-10-8
EINECS: 203-462-3
UN: 1277
ADR: 3,II
Merck Index: 14,7843

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR