Tisk…

Tributylamon 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A14868.0D A14868.0F A14868.AP
A14868.0DEA 12430 CZK
A14868.0D A14868.0F A14868.AP
Tributylamon 98%
Tributylamon

Purity: 98% CAS No.: 102-82-9 Molecular Formula: C12H27N Formula Weight: 185.36 Flash Point: 71°(159°F) Refractive Index: 1.4290 Odour: Amine-like pH: 10.6 Melting Point: -70° Boiling Point: 216° Beilstein Reference: 1698872 Merck Reference: 14,9618 UN No.: 2542 MDL No.: MFCD00009431

Vzorec: C₁₂H₂₇N
Bod varu: 216 °C
Bod tání: < –70 °C
Hustota: 0,778
Bod vzplanutí: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009431
CAS číslo: 102-82-9
EINECS: 203-058-7
UN: 2542
ADR: 6.1,II
Merck Index: 14,9618

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR