Tisk…

1,1,1-Trifluoroacetylacetone ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A14682.06 A14682.14 A14682.22
A14682.06EA 1270 CZK
A14682.06 A14682.14 A14682.22
1,1,1-Trifluoroacetylacetone ≥98%
1,1,1-Trifluoroacetylacetone

Purity: 98% CAS No.: 367-57-7 Molecular Formula: CF3COCH2COCH3 Formula Weight: 154.09 Flash Point: 26°(79°F) Refractive Index: 1.3890 Boiling Point: 105-107° Beilstein Reference: 1705177 UN No.: 1224 MDL No.: MFCD00000427

Vzorec: C₅H₅F₃O₂
Bod varu: 105…107 °C
Hustota: 1,274
Bod vzplanutí: 26 °C (79 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000427
CAS číslo: 367-57-7
EINECS: 206-698-5
UN: 1224
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR