Tisk…

5-Chloro-1-pentanol 95%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 5-Chloropentan-1-ol

L12088.09 L12088.18
L12088.09EA 3030 CZK
L12088.09 L12088.18
5-Chloro-1-pentanol 95%
5-Chloro-1-pentanol

Purity: 95% CAS No.: 5259-98-3 Molecular Formula: Cl(CH2)5OH Formula Weight: 122.60 Refractive Index: 1.4540 Boiling Point: 102-103°/8mm MDL No.: MFCD00039556

Vzorec: C₅H₁₁ClO
MW: 122,59 g/mol
Bod varu: 102…103 °C (8 mmHg)
Hustota: 1,060
Storage Temperature: Ambient
MDL Number: MFCD00039556
CAS číslo: 5259-98-3
EINECS: 226-067-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR