Chloroacetaldehyde dimethyl acetal, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-Chloro-2,2-dimethoxyethane
10906-1KG
10906-210KG
10906-25KG
SIAL10906-1KGEA
3270
CZK
SIAL10906-1KG
SIAL10906-210KG
SIAL10906-25KG
Chloroacetaldehyde dimethyl acetal, Sigma-Aldrich®
Chloroacetaldehyde dimethyl acetal
Balení: produced by Wacker Chemie AG
Vzorec:
C₄H₉ClO₂ MW: 124,57 g/mol Bod varu: 128 °C (1013 hPa) Bod tání: < –73 °C Hustota: 1,098 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000948 CAS číslo: 97-97-2 EINECS: 202-624-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...