Tisk…

5-Chloro-2-pentanone ≥94%

Dodavatel: Thermo Scientific
Varování

Synonyms: 5-Chloropentan-2-one

A10269.14 A10269.22 A10269.36
A10269.14EA 1210 CZK
A10269.14 A10269.22 A10269.36
5-Chloro-2-pentanone ≥94%
5-Chloro-2-pentanone

Purity: 94% CAS No.: 5891-21-4 Molecular Formula: C5H9ClO Formula Weight: 120.58 Storage Temperature: 2°C to 8°C Flash Point: 35°(95°F) Refractive Index: 1.4380 Boiling Point: 63-64°/12mm Beilstein Reference: 1098490 UN No.: 1224 MDL No.: MFCD00001008

Vzorec: C₅H₉ClO
MW: 120,58 g/mol
Bod varu: 63…64 °C (12 mmHg)
Bod tání: –48 °C
Hustota: 1,053
Bod vzplanutí: 35 °C (95 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00001008
CAS číslo: 5891-21-4
EINECS: 227-565-8
UN: 1224
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR