3-Chlorobutanone, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
3-Chloro-2-butanone
10904-1KG
10904-25KG
SIAL10904-1KGEA
4560
CZK
SIAL10904-1KG
SIAL10904-25KG
3-Chlorobutanone, Sigma-Aldrich®
3-Chlorobutanone
Vzorec:
C₄H₇ClO MW: 106.55 Bod varu: 114 Hustota: 1.06 Storage Temperature: Ambient |
MDL Number:
MFCD00000865 CAS číslo: 4091-39-8 EINECS: 223-834-9 UN: 1224 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...