Tisk…

3-Chlorobutanone 96%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 3-Chloro-2-butanone

L14158.22 L14158.36
L14158.22EA 1110 CZK
L14158.22 L14158.36
3-Chlorobutanone 96%
3-Chlorobutanone

Purity: 96% CAS No.: 4091-39-8 Molecular Formula: C4H7ClO Formula Weight: 106.55 Flash Point: 28°(82°F) Refractive Index: 1.4210 Melting Point: -60° Boiling Point: 113-116° Beilstein Reference: 385637 UN No.: 1224 MDL No.: MFCD00000865

Vzorec: C₄H₇ClO
MW: 106.55
Bod varu: 113…116 °C
Bod tání: < –60 °C
Hustota: 1.06
Bod vzplanutí: 28 °C (82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000865
CAS číslo: 4091-39-8
EINECS: 223-834-9
UN: 1224
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR