Tisk…

Chloroform stabilised, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: Stabilised with amylene
M-Clarity™ quality level = MQ200

372978-1L 372978-100ML 372978-2L
SIAL372978-1LEA 4110 CZK
SIAL372978-1L SIAL372978-100ML SIAL372978-2L
Chloroform stabilised, Sigma-Aldrich®
Chloroform
Vzorec: CHCl₃
Bod varu: 61,7 °C (1013 hPa)
Bod tání: –63 °C
Hustota: 1,4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS číslo: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR