Tisk…
Chloroform analytical reagent, Supelco®
Dodavatel: Merck
Nebezpečí
Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
M-Clarity™ quality level = MQ300
1.02442.1000 1.02442.2500
1.02442.1000EA 1450 CZK
1.02442.1000 1.02442.2500
Chloroform analytical reagent, Supelco®
Chloroform
Vzorec: CHCl₃
MW: 119.38
Bod varu: 61.7
Bod tání: –63 °C
Hustota: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS číslo: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM