1-Chlorohexadecane ≥97%
Dodavatel: Thermo Fisher Scientific
B23406.AE
B23406.AP
B23406.AEEA
1200
CZK
B23406.AE
B23406.AP
1-Chlorohexadecane ≥97%
1-Chlorohexadecane
Purity: 97%
CAS No.: 4860-03-1
Molecular Formula: C16H33Cl
Formula Weight: 260.89
Flash Point: 136°(276°F)
Refractive Index: 1.4490
Melting Point: 8-10°
Boiling Point: 322°
Beilstein Reference: 1748498
MDL No.: MFCD00000959
Vzorec:
C₁₆H₃₃Cl Bod varu: 322 °C Bod tání: 8…10 °C Hustota: 0,865 Bod vzplanutí: 136 °C (276 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000959 CAS číslo: 4860-03-1 EINECS: 225-461-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...