1,2,3-Trichlorpropan ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 98+%
CAS No.: 96-18-4
Molecular Formula: C3H5Cl3
Formula Weight: 147.43
Flash Point: 74°(165°F)
Refractive Index: 1.4822
Melting Point: -14°
Boiling Point: 156-158°
Beilstein Reference: 1732068
UN No.: 2810
MDL No.: MFCD00000946
Vzorec:
ClH₂CCHClCH₂Cl Bod varu: 156…158 °C Bod tání: –14 °C Hustota: 1,385 Bod vzplanutí: 74 °C (165 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000946 CAS číslo: 96-18-4 EINECS: 202-486-1 UN: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...