Phenyl acetate 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Acetic acid phenyl ester
B22561.0B
B22561.30
B22561.0BEA
3440
CZK
B22561.0B
B22561.30
Phenyl acetate 97%
Phenyl acetate
Purity: 99%
CAS No.: 122-79-2
Molecular Formula: CH3CO2C6H5
Formula Weight: 136.15
Flash Point: 76°(168°F)
Refractive Index: 1.5020
Odour: Phenol-like
Boiling Point: 195-196°
Beilstein Reference: 636458
Merck Reference: 14,7267
MDL No.: MFCD00008699
Vzorec:
C₈H₈O₂ MW: 136.15 Bod varu: 195…196 °C Hustota: 1.075 Bod vzplanutí: 76 °C (168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008699 CAS číslo: 122-79-2 EINECS: 204-575-0 Merck Index: 12,07421 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...