Octan olovičitý ≥95%, powder
Dodavatel: MP Biomedicals
|
Nebezpečí
|
0215518225
0215518290
0215518280
ICNA0215518225EA
1930
CZK
ICNA0215518225
ICNA0215518290
ICNA0215518280
Octan olovičitý ≥95%, powder
Octan olovičitý
Lead tetraacetate smoothly induces the addition of difluorodiiodomethane to alkenes and alkynes.
Vzorec:
(H₃CCOO)₄Pb Bod tání: 175…180 °C Hustota: ∼2,22 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008693 CAS číslo: 546-67-8 EINECS: 208-908-0 UN: 1616 ADR: 6.1,III Merck Index: 13,05442 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...