Tisk…

Propionan sodný ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Propionic acid sodium salt, Sodiumpropionate

A17440.0E A17440.36
A17440.0EEA 3130 CZK
A17440.0E A17440.36
Propionan sodný ≥99%
Propionan sodný

Purity: 99% CAS No.: 137-40-6 Molecular Formula: C3H5NaO2 Formula Weight: 96.06 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 285-291° Merck Reference: 14,8669 MDL No.: MFCD00002759

Vzorec: C₃H₅NaO₂
Bod tání: 285…291 °C
Bod vzplanutí: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002759
CAS číslo: 137-40-6
EINECS: 205-290-4
Merck Index: 14,8669

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR