Tisk…

Undecanoic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Hendecanoic acid, Undecylic acid

A11244.18 A11244.30 A11244.0B
A11244.18EA 1040 CZK
A11244.18 A11244.30 A11244.0B
Undecanoic acid ≥98%
Undecanoic acid

Purity: 98% CAS No.: 112-37-8 Molecular Formula: CH3(CH2)9CO2H Formula Weight: 186.30 Flash Point: 110°(230°F) Melting Point: 26-29° Boiling Point: 283-285° Beilstein Reference: 1759287 MDL No.: MFCD00002730

Vzorec: C₁₁H₂₂O₂
MW: 186.29
Bod varu: 283…285 °C
Bod tání: 26…29 °C
Hustota: 0.89
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002730
CAS číslo: 112-37-8
EINECS: 203-964-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR